Assay Details
Target Gene Details
Entrez Gene ID: | 23312 |
Gene Name: | Dmx like 2 |
Gene Aliases: |
PEPNS, RC3 |
Location: |
Chr.15:51447724-51622833 on Build GRCh38 |
Assay Gene Location: | Within Intron 35 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DMXL2 | NM_001174116.1 | NP_001167587.1 | ||
| NM_001174117.1 | NP_001167588.1 | |||
| NM_015263.3 | NP_056078.2 | |||
| XM_005254255.1 | XP_005254312.1 | |||
| XM_005254256.1 | XP_005254313.1 | |||
| XM_017022034.1 | XP_016877523.1 | |||
| AB020663.2 | BAA74879.2 | |||
| AF389880.1 | AAL93215.1 | |||
| BC022050.1 | AAH22050.1 | |||
| BC140781.1 | ||||
| BC144539.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3636461 | Chr.15:51022583 - 51482654 on Build GRCh38 | Gain |
|
| nsv471243 | Chr.15:51053288 - 51725899 on Build GRCh38 | Gain |
|
| esv2760031 | Chr.15:50990060 - 51538142 on Build GRCh38 | Gain |
|
| dgv4526n54 | Chr.15:50582767 - 51475068 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map