Assay Details
Target Gene Details
Entrez Gene ID: | 1588 |
Gene Name: | cytochrome P450 family 19 subfamily A member 1 |
Gene Aliases: |
ARO, ARO1, CPV1, CYAR, CYP19, CYPXIX, P-450AROM |
Location: |
Chr.15:51208057-51338598 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYP19A1 | NM_000103.3 | NP_000094.2 | ||
| NM_031226.2 | NP_112503.1 | |||
| XM_005254191.1 | XP_005254248.1 | |||
| AK291778.1 | ||||
| BC020767.1 | ||||
| BC022896.1 | AAH22896.1 | |||
| BC035959.1 | AAH35959.1 | |||
| BC056258.1 | AAH56258.1 | |||
| BC107785.1 | AAI07786.1 | |||
| DQ118405.1 | AAZ23558.1 | |||
| J04127.1 | AAA52132.1 | |||
| M18856.1 | AAA35556.1 | |||
| M22246.1 | AAA35557.1 | |||
| M28420.1 | AAA52141.1 | |||
| X13589.1 | CAA31929.1 | |||
| Y07508.1 | CAA68807.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3636461 | Chr.15:51022583 - 51482654 on Build GRCh38 | Gain |
|
| nsv471243 | Chr.15:51053288 - 51725899 on Build GRCh38 | Gain |
|
| nsv1047443 | Chr.15:50452584 - 51354826 on Build GRCh38 | Gain |
|
| esv34532 | Chr.15:50983931 - 51460659 on Build GRCh38 | Gain |
|
| esv2760031 | Chr.15:50990060 - 51538142 on Build GRCh38 | Gain |
|
| dgv4526n54 | Chr.15:50582767 - 51475068 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs549165695] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map