Assay Details
Target Gene Details
Entrez Gene ID: | 84952 |
Gene Name: | cingulin like 1 |
Gene Aliases: |
JACOP, PCING |
Location: |
Chr.15:57376487-57550727 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CGNL1 | NM_001252335.1 | NP_001239264.1 | ||
| NM_032866.4 | NP_116255.2 | |||
| XM_005254726.3 | XP_005254783.1 | |||
| XM_011522120.1 | XP_011520422.1 | |||
| XM_011522121.1 | XP_011520423.1 | |||
| AK056673.1 | BAB71249.1 | |||
| AK307410.1 | ||||
| AY274808.1 | AAP42073.1 | |||
| AY610514.1 | AAT37906.1 | |||
| BC030995.1 | ||||
| DA272287.1 | ||||
| DC415862.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv4556n54 | Chr.15:57330402 - 57498238 on Build GRCh38 | Gain |
|
| nsv1045611 | Chr.15:57058767 - 57778609 on Build GRCh38 | Gain |
|
| nsv833022 | Chr.15:57365214 - 57537914 on Build GRCh38 | Gain |
|
| esv3636609 | Chr.15:57350452 - 57488224 on Build GRCh38 | Gain |
|
| esv3636608 | Chr.15:57350435 - 57426987 on Build GRCh38 | Gain |
|
| nsv457166 | Chr.15:57352069 - 57483628 on Build GRCh38 | Gain |
|
| dgv2607n100 | Chr.15:57340817 - 57494394 on Build GRCh38 | Gain |
|
| esv2751536 | Chr.15:57335210 - 57496210 on Build GRCh38 | Gain |
|
| esv2761873 | Chr.15:57351455 - 57486837 on Build GRCh38 | Gain |
|
| nsv833021 | Chr.15:57203215 - 57459168 on Build GRCh38 | Gain |
|
| nsv1054547 | Chr.15:57308290 - 57685461 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map