Assay Details
Target Gene Details
Entrez Gene ID: | 1198 |
Gene Name: | CDC like kinase 3 |
Gene Aliases: |
PHCLK3, PHCLK3/152 |
Location: |
Chr.15:74598476-74645418 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLK3 | XM_017021908.1 | XP_016877397.1 | ||
| XM_017021909.1 | XP_016877398.1 | |||
| XM_017021910.1 | XP_016877399.1 | |||
| AK095335.1 |
Target Gene Details
Entrez Gene ID: | 102723750 |
Gene Name: | uncharacterized LOC102723750 |
Gene Aliases: |
- |
Location: |
Chr.15:74601853-74611502 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC102723750 | XM_017022793.1 | XP_016878282.1 | ||
| XM_017022794.1 | XP_016878283.1 | |||
| XM_017022795.1 | 7 | 1399 | XP_016878284.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv457204 | Chr.15:74592750 - 74655722 on Build GRCh38 | Loss |
|
| esv2660170 | Chr.15:74600445 - 74604158 on Build GRCh38 | Deletion |
|
| nsv817698 | Chr.15:74596822 - 74727108 on Build GRCh38 | Gain |
|
| esv3636906 | Chr.15:74600669 - 74604335 on Build GRCh38 | Loss |
|
| nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map