Assay Details
Target Gene Details
Entrez Gene ID: | 267 |
Gene Name: | autocrine motility factor receptor |
Gene Aliases: |
GP78, RNF45 |
Location: |
Chr.16:56361452-56425538 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AMFR | NM_001144.5 | NP_001135.3 | ||
| NM_001323511.1 | NP_001310440.1 | |||
| NM_001323512.1 | NP_001310441.1 | |||
| XM_005255890.3 | XP_005255947.1 | |||
| AF124145.1 | AAD56722.1 | |||
| AK023874.1 | ||||
| AK123962.1 | 1 | 499 | ||
| BC017043.1 | AAH17043.1 | |||
| BC056869.1 | AAH56869.1 | |||
| BC069197.1 | AAH69197.1 | |||
| BQ889409.1 | ||||
| L35233.1 | AAA79362.1 | |||
| M63175.1 | AAA36671.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv525929 | Chr.16:56350808 - 56367064 on Build GRCh38 | Loss |
|
| nsv833242 | Chr.16:56259354 - 56446840 on Build GRCh38 | Loss |
|
| nsv519979 | Chr.16:56306184 - 56367064 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs115787470] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map