Assay Details
Target Gene Details
Entrez Gene ID: | 11151 |
Gene Name: | coronin 1A |
Gene Aliases: |
CLABP, CLIPINA, HCORO1, IMD8, TACO, p57 |
Location: |
Chr.16:30183410-30189076 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CORO1A | NM_001193333.2 | NP_001180262.1 | ||
| NM_007074.3 | NP_009005.1 | |||
| XM_017022885.1 | XP_016878374.1 | |||
| DA256300.1 | ||||
| DC408372.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv9431 | Chr.16:30179216 - 30218525 on Build GRCh38 | Gain+Loss |
|
| nsv571819 | Chr.16:30147436 - 30204300 on Build GRCh38 | Gain |
|
| esv2758642 | Chr.16:30062634 - 30371959 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map