Assay Details
Target Gene Details
Entrez Gene ID: | 146439 |
Gene Name: | BICD family like cargo adaptor 2 |
Gene Aliases: |
BICDR-2, BICDR2, CCDC64B |
Location: |
Chr.16:3027710-3036985 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BICDL2 | NM_001103175.1 | NP_001096645.1 | ||
| XM_005255135.4 | XP_005255192.1 | |||
| AL833749.1 | 1 | 568 | CAH56245.1 | |
| BC128602.1 | ||||
| BG142147.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3637645 | Chr.16:3021600 - 3057596 on Build GRCh38 | Gain |
|
| nsv509588 | Chr.16:2919880 - 3043776 on Build GRCh38 | Insertion |
|
| nsv833125 | Chr.16:3017124 - 3131817 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map