Assay Details
Target Gene Details
Entrez Gene ID: | 57465 |
Gene Name: | TBC1 domain family member 24 |
Gene Aliases: |
DFNA65, DFNB86, DOORS, EIEE16, FIME, TLDC6 |
Location: |
Chr.16:2475104-2509669 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TBC1D24 | NM_001199107.1 | NP_001186036.1 | ||
| NM_020705.2 | NP_065756.1 | |||
| XM_017023493.1 | XP_016878982.1 | |||
| XM_017023495.1 | XP_016878984.1 | |||
| AB032997.1 | ||||
| BC112389.1 | AAI12390.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication |
|
| nsv1052260 | Chr.16:2074546 - 2629655 on Build GRCh38 | Gain |
|
| nsv833121 | Chr.16:2360975 - 2518228 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map