Assay Details
Target Gene Details
Entrez Gene ID: | 9274 |
Gene Name: | BCL tumor suppressor 7C |
Gene Aliases: |
- |
Location: |
Chr.16:30834041-30894550 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BCL7C | NM_001286526.1 | NP_001273455.1 | ||
| NM_004765.3 | NP_004756.2 | |||
| XM_011545980.2 | XP_011544282.1 | |||
| XM_017023885.1 | XP_016879374.1 | |||
| XM_017023886.1 | XP_016879375.1 | |||
| XM_017023887.1 | XP_016879376.1 | |||
| XM_017023888.1 | XP_016879377.1 | |||
| AJ223980.1 | CAA11754.1 | |||
| BC019071.1 | AAH19071.1 | |||
| BC058863.1 | AAH58863.1 |
Target Gene Details
Entrez Gene ID: | 101928736 |
Gene Name: | MIR762 host gene |
Gene Aliases: |
- |
Location: |
Chr.16:30875460-30895220 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR762HG | NR_110940.1 | |||
| NR_110941.1 | ||||
| NR_110942.1 | ||||
| BM769724.1 | ||||
| DR760839.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv457483 | Chr.16:30892358 - 31180161 on Build GRCh38 | Loss |
|
| dgv35n68 | Chr.16:30833730 - 31036730 on Build GRCh38 | Loss |
|
| nsv953802 | Chr.16:30876680 - 30999279 on Build GRCh38 | Deletion |
|
| nsv833189 | Chr.16:30808387 - 30949474 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map