Assay Details
Target Gene Details
Entrez Gene ID: | 899 |
Gene Name: | cyclin F |
Gene Aliases: |
FBX1, FBXO1 |
Location: |
Chr.16:2425438-2458858 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCNF | XM_017023822.1 | XP_016879311.1 | ||
| XM_017023823.1 | XP_016879312.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication |
|
| nsv952903 | Chr.16:2423400 - 2476599 on Build GRCh38 | Deletion |
|
| nsv1052260 | Chr.16:2074546 - 2629655 on Build GRCh38 | Gain |
|
| nsv833121 | Chr.16:2360975 - 2518228 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs78443348] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map