Assay Details
Target Gene Details
Entrez Gene ID: | 222487 |
Gene Name: | adhesion G protein-coupled receptor G3 |
Gene Aliases: |
GPR97, PB99, PGR26 |
Location: |
Chr.16:57668192-57689378 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADGRG3 | NM_001308360.1 | NP_001295289.1 | ||
| NM_170776.4 | NP_740746.4 | |||
| XM_005255842.3 | XP_005255899.1 | |||
| XM_006721170.3 | XP_006721233.1 | |||
| XM_011522951.2 | XP_011521253.1 | |||
| XM_011522953.2 | XP_011521255.1 | |||
| XM_011522954.1 | XP_011521256.1 | |||
| AB049169.1 | BAC67700.1 | |||
| AK172801.1 | BAD18774.1 | |||
| AK301258.1 | ||||
| AK310311.1 | ||||
| AY140959.1 | AAN46673.1 | |||
| BC064508.1 | AAH64508.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833246 | Chr.16:57577216 - 57747868 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map