Assay Details
Target Gene Details
Entrez Gene ID: | 4992 |
Gene Name: | olfactory receptor family 1 subfamily F member 1 |
Gene Aliases: |
OLFMF, OR16-36, OR16-37, OR16-88, OR16-89, OR16-90, OR1F10, OR1F13P, OR1F4, OR1F5, OR1F6, OR1F7, OR1F8, OR1F9, OR3-145, ORL1023 |
Location: |
Chr.16:3188195-3206559 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR1F1 | XM_011522506.2 | XP_011520808.1 | ||
| XM_011522507.2 | XP_011520809.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv471074 | Chr.16:3153034 - 3227251 on Build GRCh38 | Gain |
|
| nsv833127 | Chr.16:3152781 - 3259413 on Build GRCh38 | Gain+Loss |
|
| nsv1044147 | Chr.16:3149392 - 3392037 on Build GRCh38 | Loss |
|
| esv3637657 | Chr.16:3199849 - 3203610 on Build GRCh38 | Loss |
|
| nsv1036149 | Chr.16:3161307 - 3326824 on Build GRCh38 | Gain |
|
| nsv1713 | Chr.16:3179211 - 3212512 on Build GRCh38 | Insertion |
|
| nsv457355 | Chr.16:3115424 - 3251897 on Build GRCh38 | Gain |
|
| esv3581941 | Chr.16:3198638 - 3205624 on Build GRCh38 | Loss |
|
| nsv524873 | Chr.16:3201522 - 3204470 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map