Assay Details
Target Gene Details
Entrez Gene ID: | 342372 |
Gene Name: | polycystin 1 like 3, transient receptor potential channel interacting |
Gene Aliases: |
- |
Location: |
Chr.16:71929538-72000401 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PKD1L3 | NM_181536.1 | 17 | 2682 | NP_853514.1 |
| XM_017023201.1 | 17 | 3105 | XP_016878690.1 | |
| XM_017023202.1 | 17 | 3105 | XP_016878691.1 | |
| XM_017023203.1 | 17 | 3105 | XP_016878692.1 | |
| XM_017023204.1 | 17 | 3105 | XP_016878693.1 | |
| AY164485.1 | 17 | 2682 | AAO32798.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1055854 | Chr.16:71888241 - 72048987 on Build GRCh38 | Loss |
|
| esv3638985 | Chr.16:71931691 - 71955811 on Build GRCh38 | Loss |
|
| esv2758654 | Chr.16:71942441 - 72240448 on Build GRCh38 | Gain |
|
| esv2714683 | Chr.16:71950823 - 72151945 on Build GRCh38 | Deletion |
|
| nsv833273 | Chr.16:71937902 - 72118955 on Build GRCh38 | Loss |
|
| nsv516948 | Chr.16:71931822 - 71954829 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map