Assay Details
Target Gene Details
Entrez Gene ID: | 1201 |
Gene Name: | CLN3, battenin |
Gene Aliases: |
BTS, JNCL |
Location: |
Chr.16:28466653-28492302 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509608 | Chr.16:28449797 - 28632895 on Build GRCh38 | Insertion |
|
| esv2758640 | Chr.16:28163817 - 29139232 on Build GRCh38 | Gain+Loss |
|
| nsv1059555 | Chr.16:28373144 - 29195838 on Build GRCh38 | Gain |
|
| nsv833180 | Chr.16:28346157 - 28574165 on Build GRCh38 | Loss |
|
| nsv9415 | Chr.16:28468881 - 28498959 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs76662516] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map