Assay Details
Target Gene Details
Entrez Gene ID: | 55114 |
Gene Name: | Rho GTPase activating protein 17 |
Gene Aliases: |
MST066, MST110, MSTP038, MSTP066, MSTP110, NADRIN, PP367, PP4534, RICH-1, RICH1, WBP15 |
Location: |
Chr.16:24919384-25015601 on Build GRCh38 |
Assay Gene Location: | Within Intron 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGAP17 | NM_001006634.2 | NP_001006635.1 | ||
| NM_018054.5 | NP_060524.4 | |||
| XM_011545873.2 | XP_011544175.1 | |||
| XM_011545874.2 | XP_011544176.1 | |||
| XM_011545875.2 | XP_011544177.1 | |||
| XM_011545876.2 | XP_011544178.1 | |||
| XM_011545877.2 | XP_011544179.1 | |||
| XM_011545878.2 | XP_011544180.1 | |||
| XM_017023390.1 | XP_016878879.1 | |||
| AJ306731.1 | CAC37948.1 | |||
| AK023281.1 | BAB14506.1 | |||
| AK027567.1 | BAB55203.1 | |||
| AK094886.1 | ||||
| AK125358.1 | BAC86144.1 | |||
| AK291691.1 | ||||
| AK301055.1 | ||||
| AK301500.1 | ||||
| AL833975.1 | CAD38819.1 | |||
| BC080195.1 | AAH80195.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758638 | Chr.16:24932074 - 25175657 on Build GRCh38 | Gain+Loss |
|
| nsv833175 | Chr.16:24846570 - 25080507 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map