Assay Details
Target Gene Details
Entrez Gene ID: | 161882 |
Gene Name: | zinc finger protein, FOG family member 1 |
Gene Aliases: |
FOG, FOG1, ZC2HC11A, ZNF408, ZNF89A |
Location: |
Chr.16:88451782-88535166 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZFPM1 | XM_011522914.2 | XP_011521216.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528456 | Chr.16:88321646 - 88485962 on Build GRCh38 | Loss |
|
| dgv5293n54 | Chr.16:88387351 - 88485962 on Build GRCh38 | Loss |
|
| nsv471112 | Chr.16:88397812 - 88485962 on Build GRCh38 | Gain |
|
| esv2758669 | Chr.16:88392764 - 88543508 on Build GRCh38 | Gain |
|
| dgv171n21 | Chr.16:88382277 - 88503975 on Build GRCh38 | Loss |
|
| nsv471111 | Chr.16:88298034 - 88503975 on Build GRCh38 | Loss |
|
| nsv1061590 | Chr.16:88228010 - 88560181 on Build GRCh38 | Gain |
|
| nsv952070 | Chr.16:88257295 - 88780192 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map