Assay Details
Target Gene Details
Entrez Gene ID: | 4166 |
Gene Name: | carbohydrate sulfotransferase 6 |
Gene Aliases: |
MCDC1 |
Location: |
Chr.16:75473124-75495419 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHST6 | NM_021615.4 | NP_067628.1 | ||
| XM_005255955.4 | XP_005256012.1 | |||
| AF280086.1 | AAG48244.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv572981 | Chr.16:75487132 - 75547262 on Build GRCh38 | Gain |
|
| nsv1063718 | Chr.16:75362071 - 75574173 on Build GRCh38 | Gain |
|
| nsv1059777 | Chr.16:75489056 - 75538815 on Build GRCh38 | Loss |
|
| esv3571642 | Chr.16:75471148 - 75515756 on Build GRCh38 | Gain |
|
| esv2758657 | Chr.16:75385716 - 75543073 on Build GRCh38 | Gain |
|
| nsv525955 | Chr.16:75439434 - 75594447 on Build GRCh38 | Loss |
|
| nsv1065995 | Chr.16:75339858 - 75592129 on Build GRCh38 | Loss |
|
| nsv457521 | Chr.16:75453860 - 75570027 on Build GRCh38 | Gain |
|
| esv2714710 | Chr.16:75482503 - 75532390 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74024796] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map