Assay Details
Target Gene Details
Entrez Gene ID: | 83723 |
Gene Name: | family with sequence similarity 57 member B |
Gene Aliases: |
FP1188 |
Location: |
Chr.16:30024423-30053026 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM57B | XM_005255613.3 | 1 | 211 | XP_005255670.1 |
| XM_005255614.3 | 1 | 211 | XP_005255671.1 | |
| XM_017023751.1 | 1 | 211 | XP_016879240.1 | |
| XM_017023752.1 | 1 | 211 | XP_016879241.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833187 | Chr.16:30051939 - 30173160 on Build GRCh38 | Loss |
|
| esv3638380 | Chr.16:30042305 - 30056210 on Build GRCh38 | Loss |
|
| nsv1160384 | Chr.16:30023148 - 30178660 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map