Assay Details
Target Gene Details
Entrez Gene ID: | 100130958 |
Gene Name: | synaptonemal complex central element protein 1 like |
Gene Aliases: |
MRP2 |
Location: |
Chr.16:77199452-77213079 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SYCE1L | NM_001129979.1 | NP_001123451.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1056676 | Chr.16:77105785 - 77820951 on Build GRCh38 | Loss |
|
| nsv1055234 | Chr.16:76969529 - 77433389 on Build GRCh38 | Gain |
|
| dgv3027n100 | Chr.16:76614676 - 77688151 on Build GRCh38 | Gain |
|
| esv3582331 | Chr.16:77199692 - 77239559 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map