Assay Details
Target Gene Details
Entrez Gene ID: | 79613 |
Gene Name: | transport and golgi organization 6 homolog |
Gene Aliases: |
TMCO7 |
Location: |
Chr.16:68843547-69085182 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TANGO6 | NM_024562.1 | 2 | 244 | NP_078838.1 |
| XM_011523327.2 | 2 | 303 | XP_011521629.1 | |
| AB051533.1 | 2 | 244 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv457515 | Chr.16:68564104 - 68890598 on Build GRCh38 | Gain |
|
| nsv572910 | Chr.16:68816880 - 68972894 on Build GRCh38 | Loss |
|
| esv3638916 | Chr.16:68847574 - 68907820 on Build GRCh38 | Loss |
|

Genomic Map