Assay Details
Target Gene Details
Entrez Gene ID: | 124491 |
Gene Name: | transmembrane protein 170A |
Gene Aliases: |
TMEM170 |
Location: |
Chr.16:75443060-75464757 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM170A | NM_001304996.1 | 3 | 4575 | NP_001291925.1 |
| NM_001304997.1 | 3 | 4559 | NP_001291926.1 | |
| NM_001304998.1 | 3 | 4371 | NP_001291927.1 | |
| NM_145254.2 | 3 | 4644 | NP_660297.1 | |
| XM_011522869.2 | 4 | 4666 | XP_011521171.1 | |
| XM_017022939.1 | 3 | 4901 | XP_016878428.1 | |
| XM_017022940.1 | 4 | 4595 | XP_016878429.1 | |
| XM_017022941.1 | 3 | 4460 | XP_016878430.1 | |
| XM_017022942.1 | 3 | 4838 | XP_016878431.1 | |
| XM_017022943.1 | 3 | 4507 | XP_016878432.1 | |
| BX647432.1 | 4 | 4106 | ||
| BX648429.1 | 4 | 2987 | ||
| BX648484.1 | 3 | 4528 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1063718 | Chr.16:75362071 - 75574173 on Build GRCh38 | Gain |
|
| nsv9460 | Chr.16:75441791 - 75445652 on Build GRCh38 | Gain+Loss |
|
| esv2758657 | Chr.16:75385716 - 75543073 on Build GRCh38 | Gain |
|
| nsv521596 | Chr.16:75432798 - 75452750 on Build GRCh38 | Loss |
|
| nsv525955 | Chr.16:75439434 - 75594447 on Build GRCh38 | Loss |
|
| nsv1065995 | Chr.16:75339858 - 75592129 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map