Assay Details
Target Gene Details
Entrez Gene ID: | 79613 |
Gene Name: | transport and golgi organization 6 homolog |
Gene Aliases: |
TMCO7 |
Location: |
Chr.16:68843547-69085182 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TANGO6 | NM_024562.1 | NP_078838.1 | ||
| XM_011523327.2 | XP_011521629.1 | |||
| AB051533.1 | ||||
| AK022750.1 | BAB14224.1 | |||
| AK095333.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv457515 | Chr.16:68564104 - 68890598 on Build GRCh38 | Gain |
|
| nsv572910 | Chr.16:68816880 - 68972894 on Build GRCh38 | Loss |
|
| esv3638916 | Chr.16:68847574 - 68907820 on Build GRCh38 | Loss |
|
| esv3638917 | Chr.16:68883193 - 68904837 on Build GRCh38 | Loss |
|
| esv2751611 | Chr.16:68861851 - 69012842 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs116941332] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map