Assay Details
Target Gene Details
Entrez Gene ID: | 3883 |
Gene Name: | keratin 33A |
Gene Aliases: |
HA3I, Ha-3I, K33A, KRTHA3A, Krt1-3, hHa3-I |
Location: |
Chr.17:41345704-41350812 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRT33A | NM_004138.3 | NP_004129.2 | ||
| AJ633621.2 | CAG17613.2 | |||
| AK314088.1 | ||||
| BC069135.1 | AAH69135.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2051 | Chr.17:41348539 - 41376166 on Build GRCh38 | Deletion |
|
| dgv3171n100 | Chr.17:41346146 - 41371345 on Build GRCh38 | Loss |
|
| nsv483014 | Chr.17:41339595 - 41514244 on Build GRCh38 | Loss |
|
| nsv509656 | Chr.17:41320187 - 41384560 on Build GRCh38 | Insertion |
|
| nsv518572 | Chr.17:41318541 - 41365943 on Build GRCh38 | Loss |
|
| esv2758690 | Chr.17:41145123 - 41740591 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map