Assay Details
Target Gene Details
Entrez Gene ID: | 102723641 |
Gene Name: | HID1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.17:74970689-74975728 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HID1-AS1 | NR_110878.1 | |||
| CD515974.1 | ||||
| HY081987.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833540 | Chr.17:74838971 - 75019365 on Build GRCh38 | Loss |
|
| nsv518292 | Chr.17:74810882 - 74982818 on Build GRCh38 | Loss |
|
| nsv470601 | Chr.17:74931028 - 74982818 on Build GRCh38 | Loss |
|
| nsv576026 | Chr.17:74889500 - 74982818 on Build GRCh38 | Gain |
|
| nsv1056788 | Chr.17:74922211 - 75028689 on Build GRCh38 | Gain |
|

Genomic Map