Assay Details
Target Gene Details
Entrez Gene ID: | 644139 |
Gene Name: | phosphoinositide interacting regulator of transient receptor potential channels |
Gene Aliases: |
- |
Location: |
Chr.17:10822475-10838101 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIRT | NM_001101387.1 | 2 | 2209 | NP_001094857.1 |
| AK021549.1 | 4 | 982 | ||
| AK300493.1 | 1 | 1309 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833367 | Chr.17:10683841 - 10826633 on Build GRCh38 | Loss |
|
| nsv1058835 | Chr.17:10322963 - 10939602 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map