Assay Details
Target Gene Details
Entrez Gene ID: | 101928991 |
Gene Name: | coiled-coil domain containing 92B |
Gene Aliases: |
- |
Location: |
Chr.17:2720801-2748500 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC92B | XM_017025489.1 | XP_016880978.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1065489 | Chr.17:2631416 - 3948349 on Build GRCh38 | Gain |
|
| nsv833340 | Chr.17:2705367 - 2783502 on Build GRCh38 | Gain+Loss |
|
| nsv574225 | Chr.17:2694675 - 2764987 on Build GRCh38 | Loss |
|
| esv2660206 | Chr.17:2679367 - 3519614 on Build GRCh38 | Deletion |
|
| nsv833339 | Chr.17:2578764 - 2769775 on Build GRCh38 | Loss |
|

Genomic Map