Assay Details
Target Gene Details
Entrez Gene ID: | 100128006 |
Gene Name: | uncharacterized LOC100128006 |
Gene Aliases: |
- |
Location: |
Chr.17:12760134-12790284 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100128006 | NR_104607.1 | |||
| BX649083.1 | ||||
| DA231808.1 |
Target Gene Details
Entrez Gene ID: | 93649 |
Gene Name: | myocardin |
Gene Aliases: |
MYCD |
Location: |
Chr.17:12665890-12768949 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYOCD | NM_001146312.2 | 14 | 7635 | NP_001139784.1 |
| NM_153604.3 | 13 | 7491 | NP_705832.1 | |
| XM_005256863.1 | 13 | 7506 | XP_005256920.1 | |
| XM_005256864.1 | 15 | 7679 | XP_005256921.1 | |
| XM_017025342.1 | 13 | 7509 | XP_016880831.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34095 | Chr.17:12436261 - 12841981 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map