Assay Details
Target Gene Details
Entrez Gene ID: | 55036 |
Gene Name: | coiled-coil domain containing 40 |
Gene Aliases: |
CILD15, FAP172 |
Location: |
Chr.17:80036632-80100613 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC40 | NM_001243342.1 | NP_001230271.1 | ||
| NM_017950.3 | NP_060420.2 | |||
| XM_005257492.4 | XP_005257549.1 | |||
| XM_011524963.2 | XP_011523265.1 | |||
| XM_011524965.2 | XP_011523267.1 | |||
| XM_017024807.1 | XP_016880296.1 | |||
| AK000760.1 | BAA91365.1 | |||
| BC035251.1 | AAH35251.1 | |||
| BC058288.1 | AAH58288.1 | |||
| DA842356.1 | ||||
| DB230102.1 | ||||
| DB236682.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1066606 | Chr.17:79955666 - 80224006 on Build GRCh38 | Gain |
|
| nsv833561 | Chr.17:79977946 - 80167051 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map