Assay Details
Target Gene Details
Entrez Gene ID: | 124535 |
Gene Name: | heat shock transcription factor 5 |
Gene Aliases: |
HSF 5, HSTF 5 |
Location: |
Chr.17:58420167-58488401 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HSF5 | NM_001080439.2 | NP_001073908.2 | ||
| XM_011524283.1 | XP_011522585.1 | |||
| BC033017.1 | AAH33017.1 | |||
| BC033020.1 | AAH33020.1 | |||
| BC115385.1 | ||||
| BC115386.1 |
More Information
Set Membership: |
|

Genomic Map