Assay Details
Target Gene Details
Entrez Gene ID: | 7473 |
Gene Name: | Wnt family member 3 |
Gene Aliases: |
INT4, TETAMS |
Location: |
Chr.17:46762506-46818760 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WNT3 | NM_030753.4 | NP_110380.1 | ||
| AB067628.1 | BAB70502.1 | |||
| BC112116.1 | AAI12117.1 | |||
| BC112118.1 | AAI12119.1 | |||
| BC114219.1 | AAI14220.1 | |||
| BF515681.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833465 | Chr.17:46629130 - 46812383 on Build GRCh38 | Loss |
|
| esv2656635 | Chr.17:46204086 - 47091135 on Build GRCh38 | Deletion |
|
| esv2758692 | Chr.17:46006548 - 47199967 on Build GRCh38 | Gain+Loss |
|
| nsv428343 | Chr.17:46314456 - 46836265 on Build GRCh38 | Gain+Loss |
|
| nsv483083 | Chr.17:46654819 - 46812153 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map