Assay Details
Target Gene Details
Entrez Gene ID: | 9772 |
Gene Name: | transmembrane protein 94 |
Gene Aliases: |
KIAA0195 |
Location: |
Chr.17:75441159-75500452 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470603 | Chr.17:75468213 - 75741366 on Build GRCh38 | Loss |
|
| nsv516133 | Chr.17:75420733 - 75763223 on Build GRCh38 | Loss |
|
| nsv576027 | Chr.17:75249939 - 75750289 on Build GRCh38 | Gain |
|
| nsv833542 | Chr.17:75441043 - 75544993 on Build GRCh38 | Loss |
|

Genomic Map