Assay Details
Target Gene Details
Entrez Gene ID: | 96597 |
Gene Name: | TBC1 domain family member 27 |
Gene Aliases: |
- |
Location: |
Chr.17:16922772-16926973 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TBC1D27 | XM_017025506.1 | 5 | 762 | XP_016880995.1 |
| AK024458.1 | 14 | 3652 | BAB15748.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1055688 | Chr.17:16897474 - 16965546 on Build GRCh38 | Gain |
|
| nsv833379 | Chr.17:16780271 - 16956225 on Build GRCh38 | Gain |
|
| nsv526289 | Chr.17:16918311 - 16934051 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map