Assay Details
Target Gene Details
Entrez Gene ID: | 401884 |
Gene Name: | ADP-ribosylation factor pseudogene |
Gene Aliases: |
- |
Location: |
Chr.17:45620329-45637963 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MGC57346 | NR_026680.3 | |||
NR_027295.2 | ||||
AK311451.1 |
Target Gene Details
Entrez Gene ID: | 104909134 |
Gene Name: | MGC57346-CRHR1 readthrough |
Gene Aliases: |
- |
Location: |
Chr.17:45620329-45835828 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MGC57346-CRHR1 | NM_001256299.2 | NP_001243228.1 | ||
NM_001303016.1 | NP_001289945.1 | |||
AK295559.1 | ||||
AK297590.1 | ||||
FJ543100.1 | ||||
HY123248.1 | ||||
HY316497.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2762433 | Chr.17:45496768 - 46710828 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv3176n100 | Chr.17:45496756 - 45633658 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
esv2758691 | Chr.17:45380520 - 45738657 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1060303 | Chr.17:45597851 - 45670724 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
esv26189 | Chr.17:45496108 - 45634334 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() |
nsv833458 | Chr.17:45567820 - 45707795 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |

More Information
Additional Information:
For this assay, SNP(s) [rs73307582] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
