Assay Details
Target Gene Details
Entrez Gene ID: | 57636 |
Gene Name: | Rho GTPase activating protein 23 |
Gene Aliases: |
- |
Location: |
Chr.17:38419286-38512392 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGAP23 | XM_006721991.1 | XP_006722054.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428341 | Chr.17:37855347 - 38600766 on Build GRCh38 | Gain+Loss |
|
| nsv469701 | Chr.17:38394019 - 38545635 on Build GRCh38 | Loss |
|
| esv2758688 | Chr.17:37706608 - 38600766 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map