Assay Details
Target Gene Details
Entrez Gene ID: | 23326 |
Gene Name: | ubiquitin specific peptidase 22 |
Gene Aliases: |
USP3L |
Location: |
Chr.17:20999593-21043409 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP22 | NM_015276.1 | NP_056091.1 | ||
| XM_005256575.2 | XP_005256632.1 | |||
| AB028986.1 | ||||
| BC110499.1 | AAI10500.1 | |||
| BC126898.1 | ||||
| BX640815.1 | CAE45893.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1059146 | Chr.17:20849194 - 22546654 on Build GRCh38 | Gain |
|
| nsv827917 | Chr.17:20992787 - 21058464 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map