Assay Details
Target Gene Details
Entrez Gene ID: | 284106 |
Gene Name: | CDGSH iron sulfur domain 3 |
Gene Aliases: |
Miner2 |
Location: |
Chr.17:38730257-38735605 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CISD3 | NM_001136498.1 | NP_001129970.1 | ||
| AK097047.1 | ||||
| AK098433.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833438 | Chr.17:38593441 - 38821266 on Build GRCh38 | Loss |
|
| nsv470585 | Chr.17:38701569 - 38760346 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs77317404] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map