Assay Details
Target Gene Details
Entrez Gene ID: | 84440 |
Gene Name: | RAB11 family interacting protein 4 |
Gene Aliases: |
RAB11-FIP4 |
Location: |
Chr.17:31391624-31538217 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAB11FIP4 | NM_032932.5 | NP_116321.2 | ||
| AB058724.1 | BAB47450.1 | |||
| AJ314646.2 | CAC44535.1 | |||
| AY169244.1 | AAO21970.1 | |||
| BC093914.1 | AAH93914.1 | |||
| BC101517.1 | AAI01518.1 | |||
| DR000701.1 |
More Information
Set Membership: |
|

Genomic Map