Assay Details
Target Gene Details
Entrez Gene ID: | 3837 |
Gene Name: | karyopherin subunit beta 1 |
Gene Aliases: |
IMB1, IPO1, IPOB, Impnb, NTF97 |
Location: |
Chr.17:47649838-47683638 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KPNB1 | NM_002265.5 | NP_002256.2 | ||
| AK316421.1 | 1 | 342 | ||
| DB083013.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv525267 | Chr.17:47374528 - 47658340 on Build GRCh38 | Gain |
|
| esv2758693 | Chr.17:47443008 - 47687590 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map