Assay Details
Target Gene Details
Entrez Gene ID: | 100506736 |
Gene Name: | schlafen family member 12 like |
Gene Aliases: |
- |
Location: |
Chr.17:35474906-35537861 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLFN12L | XM_017024005.1 | XP_016879494.1 | ||
| XM_017024006.1 | XP_016879495.1 | |||
| XM_017024007.1 | XP_016879496.1 | |||
| XM_017024008.1 | 3 | 3256 | XP_016879497.1 | |
| XM_017024009.1 | 3 | 3256 | XP_016879498.1 | |
| AK172761.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3892999 | Chr.17:35252863 - 35925533 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map