Assay Details
Target Gene Details
Entrez Gene ID: | 9349 |
Gene Name: | ribosomal protein L23 |
Gene Aliases: |
L23, rpL17 |
Location: |
Chr.17:38850068-38853800 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPL23 | NM_000978.3 | NP_000969.1 | ||
| AK024749.1 | ||||
| BC003518.1 | AAH03518.1 | |||
| BC010114.2 | AAH10114.1 | |||
| BC062716.1 | AAH62716.1 | |||
| BC104651.1 | AAI04652.1 | |||
| BC106061.1 | AAI06062.1 | |||
| CA437923.1 | ||||
| X52839.1 | CAA37023.1 | |||
| X55954.1 | CAA39417.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833439 | Chr.17:38743861 - 38923753 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map