Assay Details
Target Gene Details
Entrez Gene ID: | 124535 |
Gene Name: | heat shock transcription factor 5 |
Gene Aliases: |
HSF 5, HSTF 5 |
Location: |
Chr.17:58420167-58488401 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HSF5 | NM_001080439.2 | 6 | 3784 | NP_001073908.2 |
| XM_011524283.1 | 6 | 3797 | XP_011522585.1 | |
| AL137385.1 | 1 | 1811 | ||
| BC033017.1 | 6 | 3748 | AAH33017.1 | |
| BC033020.1 | 6 | 3771 | AAH33020.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv575800 | Chr.17:58344345 - 58432504 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map