Assay Details
Target Gene Details
Entrez Gene ID: | 57690 |
Gene Name: | trinucleotide repeat containing 6C |
Gene Aliases: |
- |
Location: |
Chr.17:77974984-78108835 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TNRC6C | NM_001142640.1 | 22 | 8697 | NP_001136112.1 |
| NM_018996.3 | 21 | 8589 | NP_061869.2 | |
| XM_017024906.1 | 23 | 14349 | XP_016880395.1 | |
| XM_017024907.1 | 22 | 14241 | XP_016880396.1 |
Target Gene Details
Entrez Gene ID: | 100131096 |
Gene Name: | TNRC6C antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.17:78107401-78111799 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TNRC6C-AS1 | NR_040071.1 | 2 | 2829 | |
| AK092916.1 | 2 | 1903 | ||
| AK094898.1 | 2 | 2301 | ||
| AK127699.1 | 1 | 2080 | ||
| AY129024.1 | 1 | 2575 | AAM98767.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833554 | Chr.17:77977426 - 78151914 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map