Assay Details
Target Gene Details
Entrez Gene ID: | 7484 |
Gene Name: | Wnt family member 9B |
Gene Aliases: |
WNT14B, WNT15 |
Location: |
Chr.17:46833201-46886730 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WNT9B | XM_011525178.2 | XP_011523480.1 | ||
| AK127268.1 | BAC86908.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3554490 | Chr.17:46837108 - 46839088 on Build GRCh38 | Deletion |
|
| esv2656635 | Chr.17:46204086 - 47091135 on Build GRCh38 | Deletion |
|
| esv2758692 | Chr.17:46006548 - 47199967 on Build GRCh38 | Gain+Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs112363431,rs78389497] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map