Assay Details
Target Gene Details
Entrez Gene ID: | 105371485 |
Gene Name: | uncharacterized LOC105371485 |
Gene Aliases: |
- |
Location: |
Chr.17:2017716-2020706 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105371485 | NR_135642.1 | 2 | 1425 |
Target Gene Details
Entrez Gene ID: | 146760 |
Gene Name: | reticulon 4 receptor like 1 |
Gene Aliases: |
NGRH2, NgR3 |
Location: |
Chr.17:1934677-2025345 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RTN4RL1 | NM_178568.3 | NP_848663.1 | ||
| AL535679.3 | ||||
| BC128608.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33335 | Chr.17:1779713 - 2048672 on Build GRCh38 | Loss |
|
| nsv827857 | Chr.17:1954024 - 2060529 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map