Assay Details
Target Gene Details
Entrez Gene ID: | 9326 |
Gene Name: | zinc finger HIT-type containing 3 |
Gene Aliases: |
TRIP3 |
Location: |
Chr.17:36486627-36499310 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZNHIT3 | NM_001281432.1 | NP_001268361.1 | ||
NM_001281433.1 | NP_001268362.1 | |||
NM_001281434.1 | NP_001268363.1 | |||
NM_004773.3 | NP_004764.1 | |||
NR_104009.1 | ||||
NR_104010.1 | ||||
NR_104011.1 | ||||
AB209193.1 | 1 | 2974 | BAD92430.1 | |
AF400652.1 | AAM82423.1 | |||
AK290858.1 | ||||
AK308583.1 | ||||
AK310022.1 | ||||
BC017931.1 | AAH17931.1 | |||
BE260293.1 | ||||
BG032116.1 | ||||
BI767446.1 | ||||
CB996852.1 | ||||
HY045632.1 | ||||
L40410.1 | AAC41737.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3640472 | Chr.17:36483375 - 36567136 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
esv2763162 | Chr.17:36460442 - 36567250 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
esv3640471 | Chr.17:36461662 - 36600489 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
nsv469853 | Chr.17:36387554 - 36574537 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv471697 | Chr.17:36357257 - 36574537 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv516100 | Chr.17:36459737 - 36549567 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() |
dgv573e214 | Chr.17:36461232 - 36515958 on Build GRCh38 | Gain |
![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
