Assay Details
Target Gene Details
Entrez Gene ID: | 146779 |
Gene Name: | EF-hand calcium binding domain 3 |
Gene Aliases: |
- |
Location: |
Chr.17:62370218-62416479 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EFCAB3 | NM_001144933.1 | NP_001138405.1 | ||
| EG327376.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758701 | Chr.17:62218196 - 62395480 on Build GRCh38 | Gain+Loss |
|
| nsv833508 | Chr.17:62217320 - 62410910 on Build GRCh38 | Gain+Loss |
|
| nsv575845 | Chr.17:62334347 - 62377715 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map