Assay Details
Target Gene Details
Entrez Gene ID: | 81871 |
Gene Name: | keratin associated protein 4-6 |
Gene Aliases: |
KAP4.15, KAP4.6, KRTAP4-15, KRTAP4.15 |
Location: |
Chr.17:41139433-41140487 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRTAP4-6 | NM_030976.1 | 1 | 680 | NP_112238.1 |
| AJ406938.1 | 1 | 379 | CAC27577.1 | |
| AJ406945.1 | 1 | 644 | CAC27584.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833446 | Chr.17:41059613 - 41247007 on Build GRCh38 | Gain |
|

Genomic Map