Assay Details
Target Gene Details
Entrez Gene ID: | 125058 |
Gene Name: | TBC1 domain family member 16 |
Gene Aliases: |
- |
Location: |
Chr.17:79932343-80035875 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TBC1D16 | NM_001271844.1 | 8 | 7417 | NP_001258773.1 |
| NM_001271845.1 | 8 | 7456 | NP_001258774.1 | |
| NM_019020.3 | 12 | 8572 | NP_061893.2 | |
| XM_005257049.2 | 12 | 8583 | XP_005257106.1 | |
| XM_005257050.3 | 12 | 8586 | XP_005257107.1 | |
| XM_006721694.3 | 13 | 8425 | XP_006721757.1 | |
| XM_011524322.2 | 12 | 8547 | XP_011522624.1 | |
| XM_011524323.2 | 10 | 8497 | XP_011522625.2 | |
| XM_011524324.2 | 8 | 8041 | XP_011522626.1 | |
| XM_017024186.1 | 10 | 8324 | XP_016879675.1 | |
| XM_017024187.1 | 11 | 8564 | XP_016879676.1 | |
| XM_017024188.1 | 10 | 7704 | XP_016879677.1 | |
| AK091923.1 | 1 | 446 | ||
| AL512709.1 | 1 | 3816 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv469865 | Chr.17:79770348 - 79957778 on Build GRCh38 | Loss |
|
| nsv470615 | Chr.17:79911412 - 79935639 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map