Assay Details
Target Gene Details
Entrez Gene ID: | 9326 |
Gene Name: | zinc finger HIT-type containing 3 |
Gene Aliases: |
TRIP3 |
Location: |
Chr.17:36486627-36499310 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNHIT3 | NM_001281432.1 | NP_001268361.1 | ||
| NM_001281433.1 | NP_001268362.1 | |||
| NM_001281434.1 | NP_001268363.1 | |||
| NM_004773.3 | NP_004764.1 | |||
| NR_104009.1 | ||||
| NR_104010.1 | ||||
| NR_104011.1 | ||||
| AB209193.1 | 1 | 2036 | BAD92430.1 | |
| AF400652.1 | AAM82423.1 | |||
| AK290858.1 | ||||
| AK308583.1 | ||||
| AK310022.1 | ||||
| BC017931.1 | AAH17931.1 | |||
| BE260293.1 | ||||
| BG032116.1 | ||||
| BI767446.1 | ||||
| CB996852.1 | ||||
| HY045632.1 | ||||
| L40410.1 | AAC41737.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3640472 | Chr.17:36483375 - 36567136 on Build GRCh38 | Gain |
|
| esv2763162 | Chr.17:36460442 - 36567250 on Build GRCh38 | Gain |
|
| esv3640471 | Chr.17:36461662 - 36600489 on Build GRCh38 | Gain |
|
| nsv469853 | Chr.17:36387554 - 36574537 on Build GRCh38 | Gain+Loss |
|
| nsv471697 | Chr.17:36357257 - 36574537 on Build GRCh38 | Gain |
|
| nsv516100 | Chr.17:36459737 - 36549567 on Build GRCh38 | Gain+Loss |
|
| dgv573e214 | Chr.17:36461232 - 36515958 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map