Assay Details
Target Gene Details
Entrez Gene ID: | 253962 |
Gene Name: | CACNA1G antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.17:50556207-50562108 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CACNA1G-AS1 | NR_038439.1 | 2 | 788 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833477 | Chr.17:50516880 - 50678469 on Build GRCh38 | Loss |
|

Genomic Map